Learn More
Sino Biological Anti-Thy1/CD90 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100673T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the human CD90, and purified by antigen affinity chromatography.
Catalog No. 50-161-1790