Learn More
Sino Biological Anti-LY6K Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100681T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the human LY6K, and purified by antigen affinity chromatography.
Catalog No. 50-161-1805