Learn More
Sino Biological Anti-HMGA2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100686T34100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human HMGA2, and purified by antigen affinity chromatography.
Catalog No. 50-161-1811