Learn More
Sino Biological Anti-Aggrecan/ACAN Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100693T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human ACAN, and purified by antigen affinity chromatography.
Catalog No. 50-161-1823