Learn More
Sino Biological Recombinant Human AGRP Protein (hFc Tag)
Supplier:  Sino Biological 10070H01H100
Encompass_Preferred
A DNA sequence encoding the human AGRP (NP_001129.1) (Ser85-Thr132) was expressed with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-161-3935