Learn More
Sino Biological Anti-Claudin-4/CLDN4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100707T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the human CLDN4, and purified by antigen affinity chromatography.
Catalog No. 50-161-1855