Learn More
Sino Biological Anti-Nucleostemin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100716T34100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human GNL3 / Nucleostemin, and purified by antigen affinity chromatography.
Catalog No. 50-161-1870