Learn More
Sino Biological Recombinant Human CD14 Protein (hFc Tag)
Supplier:  Sino Biological 10073H01H100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Thr 20-Cys 352) of the mature form of human CD14 (NP_000582.1) was fused to the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-161-3942