Learn More
Sino Biological Recombinant Human CD14 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10073H08H100
Encompass_Preferred
A DNA sequence encoding the mature form of human CD14 (NP_000582.1) (Met 1-Met 344) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-3944