Learn More
Sino Biological Anti-CD14 Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 10073MM06P25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD14 (rh CD14; Catalog#10073-H08H; NP_000582.1; Met1-Met344) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-161-1929