Learn More
Sino Biological Recombinant Anti-CD14 Antibody (FITC), Rabbit Monoclonal
Supplier:  Sino Biological 10073R002F25
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD14 (rh CD14; Catalog#10073-H08H; NP_000582.1; Met1-Met344) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-161-1942