Learn More
Sino Biological Anti-CD14 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10073RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CD14 (rh CD14; Catalog#10073-H08H; NP_000582.1; Met1-Met344). CD14 specific IgG was purified by Human CD14 affinity chromatography.
Catalog No. 50-161-1950