Learn More
Sino Biological Anti-Glut4/SLC2A4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100733T42100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human SLC2A4, and purified by antigen affinity chromatography.
Catalog No. 50-161-1903