Learn More
Sino Biological Anti-MuRF1/TRIM63 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100739T34100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the human TRIM63, and purified by antigen affinity chromatography.
Catalog No. 50-161-1915