Learn More
Selleck Chemical LLC 10074-G5 10mg 413611-93-5
SDP
Supplier:  Selleck Chemical LLC S842610MG
Encompass_Preferred
10074-G5 is a c-Myc inhibitor that binds to and distorts the bHLH-ZIP domain of c-Myc (Kd = 2.8 ¿M), thereby inhibiting c-Myc/Max heterodimer formation and inhibiting its transcriptional activity (IC50 = 146 ¿M). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6050