Learn More
Medchemexpress LLC HY-100996 100mg Medchemexpress, 10074-G5 CAS:413611-93-5 Purity:98%
SDP
Catalog No. 501636876
Encompass_Preferred
Medchemexpress,HY-100996 100mg 10074-G5 CAS:413611-93-5 Formula:C18H12N4O3 10074-G5 is an inhibitor of c-Myc-Max dimerization with an IC50 of 146 μM. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-163-6876 Supplier Medchemexpress LLC Supplier No. HY100996100MG
Medchemexpress,HY-100996 100mg 10074-G5 CAS:413611-93-5 Formula:C18H12N4O3 10074-G5 is an inhibitor of c-Myc-Max dimerization with an IC50 of 146 μM. Purity:98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.