Learn More
Sino Biological Anti-ZIC3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100740T42100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ZIC3, and purified by antigen affinity chromatography.
Catalog No. 50-161-1953