Learn More
Sino Biological Anti-BMPR1A/ALK-3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100743T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human ALK-3 / BMPR1A, and purified by antigen affinity chromatography.
Catalog No. 50-161-1962