Learn More
Sino Biological Anti-CAMKIV Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100744T34100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human CAMKIV/CAMK4 , and purified by antigen affinity chromatography.
Catalog No. 50-161-1965