Learn More
eMolecules​ 10075-48-6 | 5-bromo-3-methyl-1H-indole | Synthonix | MFCD04038746 | 210.074 | C9H8BrN | 97.000 | Cc1c[nH]c2ccc(Br)cc12 | 25g | 294837928
Catalog No. 5000492097 Shop All eMolecules Inc. Products
Encompass_Preferred
5-bromo-3-methyl-1H-indole | Synthonix | 10075-48-6 | MFCD04038746 | 210.074 | C9H8BrN | 97.000 | Cc1c[nH]c2ccc(Br)cc12 | 25g | 294837928
Catalog No. 50-004-92097 Supplier eMolecules​ Supplier No. 294837928
5-bromo-3-methyl-1H-indole | Synthonix | 10075-48-6 | MFCD04038746 | 210.074 | C9H8BrN | 97.000 | Cc1c[nH]c2ccc(Br)cc12 | 25g | 294837928