Learn More
Sino Biological Anti-Cardiac Troponin I/cTnI Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100758T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human TNNI3, and purified by antigen affinity chromatography.
Catalog No. 50-161-2004