Learn More
Sino Biological Anti-CBP/CREBBP Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100760T02100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human CREBBP, and purified by antigen affinity chromatography.
Catalog No. 50-161-2007