Learn More
Sino Biological Anti-HP1 Gamma/CBX3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100766T38100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human CBX3, and purified by antigen affinity chromatography.
Catalog No. 50-161-2025