Learn More
Sino Biological Recombinant Anti-CNDP1 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10077R001100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CNDP1 (rh CNDP1; Catalog#10077-H08H; NP_116038.4; Ser27-His507).
Catalog No. 50-161-2075