Learn More
Sino Biological Anti-HDAC2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100782T46100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human HDAC2, and purified by antigen affinity chromatography.
Catalog No. 50-161-2086