Learn More
Sino Biological Anti-DNMT3A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100785T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human DNMT3A, and purified by antigen affinity chromatography.
Catalog No. 50-161-2092