Learn More
Sino Biological Anti-HP1 Alpha/CBX5 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100788T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human CBX5, and purified by antigen affinity chromatography.
Catalog No. 50-161-2104