Learn More
Sino Biological Anti-APE1/APEX1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100801T46100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human APEX1 / AP / APEx / Ref-1, and purified by antigen affinity chromatography.
Catalog No. 50-161-2125