Learn More
Sino Biological Anti-CDK1/CDC2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100802T38100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human CDC2 Kinase / CDK1, and purified by antigen affinity chromatography.
Catalog No. 50-161-2128