Learn More
Sino Biological Anti-APOBEC2 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100810T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the human APOBEC2, and purified by antigen affinity chromatography.
Catalog No. 50-161-2149