Learn More
Sino Biological Recombinant Human MMP-2 Protein, HPLC-verified
Supplier:  Sino Biological 10082HNAH100
Encompass_Preferred
A DNA sequence encoding the native human MMP2 (NP_004521.1) (Met 1-Cys 660) was expressed and purified.
Catalog No. 50-161-3957