Learn More
Sino Biological Anti-MMP-2 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10082MM05100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human MMP-2 (rh MMP-2; Catalog#10082-HNAH; NP_004521.1; Met 1-Cys660). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-161-2176