Learn More
Sino Biological Anti-Angiogenin Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100835T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Rat Angiogenin / RNASE5, and purified by antigen affinity chromatography.
Catalog No. 50-161-2195