Learn More
Sino Biological Recombinant Human PD-L1/B7-H1 Protein (ECD, mFc Tag), HPLC-verified
Supplier:  Sino Biological 10084H05H100
Encompass_Preferred
A DNA sequence encoding the human CD274 (NP_054862.1) (Met1-Thr239) was expressed with the Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-161-3963