Learn More
Sino Biological Anti-PD-L1/B7-H1 Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 10084MM36F25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PD-L1/B7-H1/CD274 (rh PD-L1/B7-H1/CD274; Catalog#10084-H08H; NP_054862.1; Met1-Thr239) and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-161-2243