Learn More
Sino Biological Recombinant Anti-PD-L1/B7-H1 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10084R015100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human PD-L1 / B7-H1 / CD274 (rh PD-L1 / B7-H1 / CD274; Catalog#10084-H08H; NP_054862.1; Met 1-Thr 239).
Catalog No. 50-161-2251