Learn More
Sino Biological Anti-PD-L1/B7-H1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10084T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human PD-L1 / B7-H1 / CD274 (rh PD-L1 / B7-H1 / CD274; Catalog#10084-H08H; NP_054862.1; Met1-Thr239). PD-L1 / B7-H1 / CD274 specific IgG was purified by Human PD-L1 / B7-H1 / CD274 affinity chromatography.
Catalog No. 50-161-2277