Learn More
Sino Biological Anti-ATF1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100841T10100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human ATF1/Protein TREB36, and purified by antigen affinity chromatography.
Catalog No. 50-161-2213