Learn More
Sino Biological Anti-SMARCC1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100848T42100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human SMARCC1, and purified by antigen affinity chromatography.
Catalog No. 50-161-2222