Learn More
Sino Biological Anti-TRAF4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100852T14100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human Traf4, and purified by antigen affinity chromatography.
Catalog No. 50-161-2280