Learn More
Sino Biological Anti-CD45 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10086MM05100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CD45 (rh CD45; Catalog#10086-H02H; NP_563578.2; Met1-Lys416). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-161-2304