Learn More
eMolecules​ 1008773-79-2 | cis-3-(tert-butoxycarbonylamino)cyclobutanecarboxylic acid | Ambeed | MFCD06657680 | 215.249 | C10H17NO4 | 97.000 | CC(C)(C)OC(=O)N[C@H]1C[C@H](C1)C(O)=O | 5g | 525009850
Catalog No. 5000482505 Shop All eMolecules Inc. Products
Encompass_Preferred
cis-3-(tert-butoxycarbonylamino)cyclobutanecarboxylic acid | Ambeed | 1008773-79-2 | MFCD06657680 | 215.249 | C10H17NO4 | 97.000 | CC(C)(C)OC(=O)N[C@H]1C[C@H](C1)C(O)=O | 5g | 525009850
Catalog No. 50-004-82505 Supplier eMolecules​ Supplier No. 525009850
cis-3-(tert-butoxycarbonylamino)cyclobutanecarboxylic acid | Ambeed | 1008773-79-2 | MFCD06657680 | 215.249 | C10H17NO4 | 97.000 | CC(C)(C)OC(=O)N[C@H]1C[C@H](C1)C(O)=O | 5g | 525009850