Learn More
Sino Biological Recombinant Human Glypican 3/GPC3 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 10088H08H100
Encompass_Preferred
A DNA sequence encoding the human GPC3 (P51654-1) (Met1-His559) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-3972