Learn More
Sino Biological Anti-FXR1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100921T02100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human FXR1, and purified by antigen affinity chromatography.
Catalog No. 50-161-2388