Learn More
Sino Biological Anti-GATAD2A Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100924T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human GATAD2A / Hp66alpha, and purified by antigen affinity chromatography.
Catalog No. 50-161-2400