Learn More
Sino Biological Recombinant Human GAPDH Protein (His Tag)
Supplier:  Sino Biological 10094H07E100
Encompass_Preferred
A DNA sequence encoding the human GAPDH (P04406) (Met 1-Glu 335) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-161-3977