Learn More
Sino Biological Anti-BCAS3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100943T34100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human BCAS3, and purified by antigen affinity chromatography.
Catalog No. 50-161-2433