Learn More
Sino Biological Anti-CADM4 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100945T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human CADM4 / IGSF4C / NECL-4, and purified by antigen affinity chromatography.
Catalog No. 50-161-2436