Learn More
Sino Biological Anti-MEK3/MAP2K3 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100952T46100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human MAP2K3/MKK3, and purified by antigen affinity chromatography.
Catalog No. 50-161-2466