Learn More
Sino Biological Anti-MEK5/MAP2K5 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 100953T10100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human MAP2K5, and purified by antigen affinity chromatography.
Catalog No. 50-161-2469